Learn More
Medchemexpress LLC Ribitol-2-13C | 488-81-3 | 98.0% | 153.14 g/mol | C4(13C)H12O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100582S110MG
Encompass_Preferred
Ribitol-2-13C is a 13C-labeled ribitol (adonitol) with the isotope at the 2-position, supplied as a crystalline pentitol for use as an internal standard and tracer in metabolomics and analytical chemistry. Available in small vial sizes for low-consumption analytical workflows, it supports accurate quantitation and structural studies.
  • Isotopically labeled at the 2-position for accurate tracing and quantitation.
  • Crystalline solid form for easy handling and precise weighing.
  • High purity (98.0%) to minimize analytical interference.
  • Suitable for LC-MS, GC-MS, and NMR workflows.
  • Small vial sizes conserve material for low-volume experiments.
  • Consistent molecular weight for reliable calculations.
Catalog No. 50-002-14826