Learn More
Medchemexpress LLC Ribitol-2-13C | 98.0% | 153.14 g·mol⁻1 | C4(13C)H12O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY100582S150MG
Encompass_Preferred
Ribitol-2-13C is a stable isotope labeled ribitol supplied as a powder for research use as a tracer and internal standard in quantitative analyses by NMR, GC-MS, and LC-MS. The material is provided with a datasheet and certificate of analysis documenting isotopic labeling and analytical purity.
  • Stable 13C label at carbon-2 for tracer and metabolic studies.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Provided as a powder for straightforward weighing and solution preparation.
  • Analytical purity reported by COA: 98.0% (NMR).
  • Molecular weight 153.14 g·mol⁻1; formula C4(13C)H12O5.
Catalog No. 50-002-13741