Learn More
Medchemexpress LLC Ribitol-2-13C (Adonitol-2-13C) | 488-81-3 | 98.0% | 153.14 g/mol | C413CH12O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY100582S125MG
Encompass_Preferred
Ribitol-2-13C is a 13C-labelled ribitol provided as a stable isotope tracer for metabolic and biochemical studies, including flux analysis of the pentose phosphate pathway and quantitation by mass spectrometry.
  • 13C label at the 2-position.
  • Stable isotope tracer for metabolic flux analysis.
  • High purity: 98.0%.
  • Supplied as a 25 mg powder for small-scale experiments.
  • Suitable for mass spectrometry and tracer quantitation workflows.
Catalog No. 50-002-13883