Learn More
Medchemexpress LLC Ribitol-3-13C | 488-81-3 | 99.7% | 153.14 g/mol | C5H12O5 (3-13C) | 25 MG
SDP
Supplier:  Medchemexpress LLC HY100582S225MG
Encompass_Preferred
Ribitol-3-13C is a stable isotope-labeled ribitol with a 13C substitution at the 3-position, provided as a high-purity crystalline material for use in isotope tracing and analytical applications. It is intended for use as an internal standard, tracer, or reference material in metabolomics and mass spectrometry workflows.
  • Stable isotope label at carbon-3 for precise tracer studies.
  • High purity suitable for mass spectrometry and quantitative analyses.
  • Solid crystalline form for reliable handling and storage.
  • Suitable as an internal standard for metabolomics workflows.
  • Compatible with common sample preparation and derivatization methods.
Catalog No. 50-002-14720