Learn More
Medchemexpress LLC Ribitol | 488-81-3 | MFCD00064291 | 98.0% | 152.15 g/mol | C5H12O5 | 10 G
SDP
Supplier:  Medchemexpress LLC HY10058210G
Encompass_Preferred
Ribitol is a crystalline pentose alcohol (the reduced form of ribose) supplied as an analytical standard for research and analytical applications. It is commonly used as an internal standard and reference compound in metabolomics, chromatography, and biochemical assays, and is provided as a bench-top reagent for laboratory use.
  • Analytical standard suitable for research and analytical applications.
  • High purity: 98.0%.
  • Molecular weight 152.15 g/mol and formula C5H12O5.
  • White crystalline solid, soluble in water for preparation of standard solutions.
  • Available in laboratory-scale pack sizes for routine use.
Catalog No. 50-002-12663