Learn More
Medchemexpress LLC Riboflavin-13C4,15N2 (vitamin B2-13C4,15N2) | 1217461-14-7 | 97.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0456S1MG
Encompass_Preferred
Riboflavin-13C4,15N2 is the 13C-labeled and 15N-labeled Riboflavin. Riboflavin is an easily absorbed micronutrient with a key role in maintaining health in humans and other animals.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • For research use only.
Catalog No. 50-004-08209