Learn More
Apexbio Technology LLC Riboflavin 83-88-5 1g
SDP
Supplier:  Apexbio Technology LLC B18241000
Encompass_Preferred
Riboflavin (83-88-5) is a water-soluble vitamin serving as the precursor for flavin adenine dinucleotide (FAD) and flavin mononucleotide (FMN) coenzymes integral to cellular oxidation-reduction reactions Riboflavin-derived FMN and FAD mediate electron transfer processes facilitating enzymatic reactions in mitochondrial energy metabolism amino acid oxidation and lipid catabolism Riboflavin exerts its biological activity primarily through facilitating redox enzyme catalysis involving flavin-dependent enzymes In cell-based studies effects of riboflavin on target enzymes commonly report an IC50 within the low micromolar to sub-millimolar concentration range depending on experimental context and model system Based on these pharmacological properties riboflavin holds research potential in elucidating cellular redox mechanisms enzymatic catalysis and metabolic pathway regulation
Catalog No. 50-224-0309