Learn More
Sigma Aldrich Fine Chemicals Biosciences Riboflavin 5-monophosphate5MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F83995MG
Encompass
Riboflavin 5AE-monophosphate (fMN) is highly soluble in water. flavins are considered as blue-light absorbing dyes.
Catalog No. 50-290-4663