Learn More
Cayman Chemical RIboflavIn 5-monphosphate 10g
SDP
Supplier:  Cayman Chemical 1816710 G
Encompass_Preferred
A coenzyme that is tightly bound to enzymes catalyzing oxidation and reduction reactions in a variety of biosynthetic pathways; binds the FMN riboswitch (RFN element) on RNA to alter gene regulation; a substrate of FMN phosphohydrolases
Catalog No. 50-296-6790