Learn More
Sigma Aldrich Fine Chemicals Biosciences Riboflavin 5-phosphate sod10G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences R777410G
Encompass
Riboflavin 5AE-phosphate (flavin mononucleotide (fMN)) is a vitamin B12 derivative containing isoalloxazine ring system.
Catalog No. 50-290-0033