Learn More
Medchemexpress LLC Riboflavin | 83-88-5 | 99.89% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB045610G
Encompass_Preferred
Riboflavin, also known as Vitamin B2, is an orally active and easily absorbed micronutrient. It acts as a precursor to flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD), which are crucial coenzymes for various enzymatic reactions. Riboflavin performs key metabolic functions by mediating the transfer of electrons in biological oxidation-reduction processes.
  • Acts as a precursor to FMN and FAD
  • Functions as a coenzyme in enzymatic reactions
  • Mediates electron transfer in biological oxidation-reduction reactions
  • Orally active and easily absorbed
  • Suitable for in vitro and in vivo studies
Catalog No. 50-004-06327