Learn More
Medchemexpress LLC Riboflavin | 83-88-5 | 99.89% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB0456
Encompass_Preferred
Riboflavin, also known as Vitamin B2 or E101, is an orally active and readily absorbed micronutrient. It acts as a precursor to flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD), which serve as crucial coenzymes for numerous enzymatic reactions and play a vital role in metabolic functions by mediating electron transfer in biological oxidation-reduction reactions.
  • Demonstrates a potent cytotoxic effect on HL60 cells after UV irradiation.
  • Enhances cell viability and glutathione reductase activity in HepG2 cells.
  • Promotes growth in Swiss albino mice.
  • Produces antinociceptive, antihyperalgesic, and anti-inflammatory effects in various rat models.
  • Inhibits the nociceptive response in mice experiencing acetic acid-induced constriction.
Catalog No. 50-004-07443