Learn More
Sigma Aldrich Fine Chemicals Biosciences Riboflavin Binding Protein10MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences R525810MG
Encompass
Riboflavin binding protein (RBP) transports riboflavin through the bloodstream of the laying hen into the eggs where the vitamin is essential for embryonic growth and development. RBP consists of a single polypeptide chain containing 9 disulfide bonds. It is a glycoprotein containing ~14% carbohydrate with sugar moieties including galactose mannose and glucosamine and is phosphorylated.
Catalog No. 50-290-1143