Learn More
Apexbio Technology LLC Ribonic acid gamma lactone 5336-08-3 100mg
SDP
Supplier:  Apexbio Technology LLC B7913100
Encompass_Preferred
Ribonic acid gamma lactone (CAS 5336-08-3) is a small-molecule inhibitor targeting -galactosidase derived from Escherichia coli It is designed to inhibit enzyme activity thereby interfering with substrate hydrolysis in carbohydrate metabolic pathways Ribonic acid gamma lactone exerts its biological activity primarily through competitive binding to the active site of -galactosidase In in vitro studies ribonic acid gamma lactone demonstrates inhibitory activity with an inhibitory constant (Ki) of approximately 26 mM Based on these pharmacological properties ribonic acid gamma lactone holds research potential in studies examining enzyme-substrate interactions and carbohydrate metabolism mediated by bacterial -galactosidase
Catalog No. 50-000-91453