Learn More
Crescent Chemical Co Inc RIBONUCLEASE A FRM BOVINE PANC
SDP
Supplier:  Crescent Chemical Co Inc SE34390.03
Encompass_Preferred
Ribonuclease A from bovine pancreas min. 70 Kunitz units/mg lyophil. RNase A content approx. 70 %.DNase-free RNase: To prepare RNase A free of DNase dissolve RNase A in TE buffer at 1 mg/ml and boil 10 to 30 minutes. Store aliquots at -20 °C to prevent microbial growth. Size - 1G Storage Condition
Catalog No. 50-248-103