Learn More
Medchemexpress LLC Ribonuclease T2 | >95.0% | 50 UG

Supplier: Medchemexpress LLC HYP7600650UG
Recombinant human Ribonuclease T2 produced in Sf9 insect cells and purified with a C-terminal His tag. Supplied as a filtered, buffer-formulated solution for biochemical characterization and in vitro research applications; store frozen and avoid repeated freeze-thaw cycles.
- Expressed in Sf9 insect cells
- C-terminal His tag for purification and detection
- Purity >95.0% by reducing SDS-PAGE
- Molecular weight approximately 35 kDa
- Supplied in 20 mM Tris, 500 mM NaCl, pH 7.4, 10% glycerol
- Supplied as a 0.22 μm filtered solution
- Store at -80°C; stable at -20°C for short-term storage; avoid repeated freeze-thaw
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.