Learn More
Medchemexpress LLC Ribonucleic acid (yeast) | 63231-63-0 | MFCD00132195 | 92.4% | 500 MG

Supplier: Medchemexpress LLC HYW250124500MG
Ribonucleic acid (yeast) is a yeast-derived RNA reagent supplied as a solid preparation for use as a biological material or an organic compound in life-science research. It is used in biochemical assays and molecular biology workflows that require bulk RNA. The material is provided in a 500 mg package and corresponds to CAS 63231-63-0.
- Solid, light yellow to brown appearance.
- High assay purity (92.4%).
- Suitable for biochemical assays and molecular biology applications.
- Store protected from light; short-term storage at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
- Supplied as a 500 MG quantity for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.