Learn More
Medchemexpress LLC Ribostamycin | 25546-65-0 | 99.9% | C17H34N4O10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY142127100MG
Encompass_Preferred
Ribostamycin (Vistamycin) is a broad-spectrum aminoglycoside antibiotic effective against both Gram-Negative and Gram-Positive bacterial infections. It also inhibits the chaperone activity of Protein Disulfide Isomerase (PDI). It has been shown to inhibit *Borrelia burgdorferi* and *Escherichia coli* strains in vitro. Studies in rats indicated that ribostamycin causes little nephrotoxicity at a dosage of 40 mg/kg via intramuscular injection daily for 14 days.
  • Effective against Gram-Negative and Gram-Positive bacterial infections.
  • Inhibits the chaperone activity of PDI.
  • Demonstrates *in vitro* activity against *Borrelia burgdorferi*.
  • Inhibits *Escherichia coli* strains.
  • Shows low nephrotoxicity in rat studies.
Catalog No. 50-003-53690