Learn More
Medchemexpress LLC Ricasetron | 117086-68-7 | MFCD00866971 | 98.6% | 313.44 g/mol | C19H27N3O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1017095MG
Encompass_Preferred
Ricasetron is a selective, orally active 5-HT3 receptor antagonist used in preclinical research to study serotonin receptor pharmacology. It has demonstrated antiemetic and anxiolytic activities and is supplied as a high-purity research chemical for biochemical and pharmacological assays.
  • High purity suitable for research (98.6%).
  • Molecular formula C19H27N3O; molecular weight 313.44 g/mol.
  • Selective 5-HT3 receptor antagonist for pharmacology and preclinical studies.
  • Useful as a reference compound in assays investigating antiemetic and anxiolytic effects.
  • Available in small quantities appropriate for laboratory experiments.
Catalog No. 50-002-48686