Learn More
Medchemexpress LLC Ricinoleic acid | 141-22-0 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN60705G
Encompass_Preferred
Ricinoleic acid is an analytical standard intended for research and analytical applications. It is a hydroxy fatty acid that serves as a feedstock for producing high-performance lubricants, cosmetics, polymers, surfactants, and coatings. This compound is the main active ingredient in Castor oil.
  • Acts as an agonist for the prostaglandin EP3 receptor (EC50 in MEG-01 is 0.5 μM), causing laxative effects and uterine contraction
  • Exhibits antianxiety-like, anti-inflammatory, and pro-inflammatory properties
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS
Catalog No. 50-004-18234