Learn More
Cayman Chemical Ricinoleic Acid methyl ester
SDP
Supplier:  Cayman Chemical 10009729500 MG
Encompass_Preferred
Ricinoleic acid methyl ester is a neutral, more lipophilic form of the free acid that can be used as an analytical standard for ricinoleic acid.
Catalog No. 50-289-0168