Learn More
Apexbio Technology LLC Ridaforolimus (Deforolimus, MK-8669) 572924-54-0 10mg
SDP
Supplier:  Apexbio Technology LLC B163910
Encompass_Preferred
Ridaforolimus (Deforolimus MK-8669) (CAS 572924-54-0) is a small-molecule inhibitor targeting the mammalian target of rapamycin (mTOR) It is designed to inhibit mTOR activity thereby modulating mTOR-mediated signal transduction pathways Ridaforolimus exerts its biological activity primarily through inhibition of phosphorylation of downstream effectors such as S6 and 4E-BP1 In cell-based studies ridaforolimus demonstrates inhibitory activity with an IC50 value of approximately 0 2 nM in HT-1080 cells Based on these pharmacological properties ridaforolimus holds research potential in oncology therapeutics development
Catalog No. 50-224-0124