Learn More
Medchemexpress LLC RIDR-PI-103 | 2581114-71-6 | 98.0% | C27H25N7O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14487610MM/1ML
Encompass_Preferred
RIDR-PI-103 is a reactive oxygen species (ROS)-induced agent release proagent featuring a self-cyclizing moiety linked to a pan-PI3K inhibitor. It demonstrates a synergistic effect with Doxorubicin in MDA-MB-361 and MDA-MB-231 cells, inhibiting cancer cell proliferation. This compound has a molecular weight of 511.53 and appears as a light yellow to yellow solid.
  • Reactive oxygen species (ROS)-induced agent release proagent
  • Features a self-cyclizing moiety
  • Linked to a pan-PI3K inhibitor
  • Synergistic effect with Doxorubicin in certain cancer cells
  • Inhibits cancer cell proliferation
  • For research use only, not for human use
Catalog No. 50-003-55879