Learn More
Medchemexpress LLC Rifabutin-d7 | 2747918-39-2 | 98.4% | 854.05 | 500 UG
SDP
Supplier:  Medchemexpress LLC HY17025S500UG
Encompass_Preferred
Rifabutin-d7 is the deuterium labeled Rifabutin, a semisynthetic ansamycin antibiotic with potent antimycobacterial properties, which inhibits DNA-dependent RNA polymerase. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during the drug development process
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-86069