Learn More
Medchemexpress LLC Rifampicin | 13292-46-1 | 98.2% | 5 G
SDP
Supplier:  Medchemexpress LLC 5GHYB0272
Encompass_Preferred
Rifampicin (Rifampin; Rifamycin AMP) is a potent, broad-spectrum antibiotic effective against bacterial pathogens. It also exhibits anti-influenza virus and anti-orthopoxvirus activities, making it suitable for a variety of research applications. This product is intended for research use only.
  • Potent, broad-spectrum antibiotic
  • Effective against bacterial pathogens
  • Exhibits anti-influenza virus activity
  • Exhibits anti-orthopoxvirus activity
Catalog No. 50-003-64453