Learn More
Gold Biotechnology Inc Rifampicin, Molecular Biology Grade 10 g
SDP
Supplier:  Gold Biotechnology Inc R12010
Encompass_Preferred
Rifampicin is a broad-spectrum antibiotic known for its efficacy against tuberculosis and other infections caused by Mycobacterium species. This semisynthetic antibiotic has also been shown to work against methicillin-resistant Staphylococcus aureus (MRSA), to activate nuclear pregnane x receptor (PXR), and to promote CYP2C-mediated metabolism.
Catalog No. 50-153-2831