Learn More
U.S. Pharmacopeia Rifampin, 13292-46-1, MFCD00151389, 300mg
Supplier:  U.S. Pharmacopeia 1604009
Encompass
Molecular formula C43H58N4O12, Molecular weight 822.94, Melting Point 361.4 - 370.4 °F (183 - 188 °C), Synonyms: Rifampicin * Rifamycin AMP
Catalog No. NC9022034