Learn More
AdipoGen Rifamycin M14
SDP
Supplier:  AdipoGen AGCN20332M010
Encompass_Preferred
Chemical. CAS 2750-76-7. Formula C43H58N2O13. MW 810.9. Semisynthetic. Ansamycin antibiotic. Selective inhibitor of bacterial DNA-dependent RNA polymerase RNAP. Effective against mycobacteria, and are therefore used in research of tuberculosis, leprosy and mycobacterium avium complex MAC infections.
Catalog No. 50-256-2579