Learn More
Apexbio Technology LLC Rifamycin sodium salt 14897-39-3 25mg
SDP
Supplier:  Apexbio Technology LLC C649825
Encompass_Preferred
Rifamycin sodium salt (14897-39-3) is a small-molecule inhibitor targeting bacterial RNA polymerase It is designed to inhibit RNA synthesis thereby suppressing bacterial growth Rifamycin sodium salt exerts its biological activity primarily through inhibition of bacterial RNA polymerase In cellular assays evaluating antibacterial potency Rifamycin sodium salt demonstrates inhibitory activity with IC50 values typically in the micromolar to submicromolar concentration range depending on bacterial strains and experimental conditions Based on these pharmacological properties Rifamycin sodium salt holds research potential in studies of bacterial transcription mechanisms antibiotic resistance pathways and the screening of antimicrobial compounds
Catalog No. 50-000-90091