Learn More
Selleck Chemical LLC Rifapentine 25mg 61379-65-5 MDL473
SDP
Supplier:  Selleck Chemical LLC S176025MG
Encompass_Preferred
Rifapentine (MDL473) is an antibiotic, which inhibits DNA-dependent RNA polymerase activity, used to treat tuberculosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-613-5