Learn More
Medchemexpress LLC HY-107902 5mg Medchemexpress, RIG-1 modulator 1 CAS:1428729-63-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1079025MG
Encompass_Preferred
Medchemexpress, HY-107902 5mg RIG-1 modulator 1 CAS:1428729-63-8 RIG-1 modulator 1 is an anti-viral compound which can be useful for the treatment of viral infections including influenza virus, HBV, HCV and HIV extracted from patent WO 2015172099 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8479