Learn More
Medchemexpress LLC HY-12037A 10mg , Rigosertib CAS:592542-59-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12037A10MG
Encompass_Preferred
HY-12037A 10mg Rigosertib CAS: 592542-59-1 Rigosertib (ON-01910) is a multi-kinase inhibitor and a selective anti-cancer agent, which induces apoptosis by inhibition the PI3 kinase/Akt pathway, promots the phosphorylation of histone H2AX and induces G2/M arrest in cell cycle. Rigosertib is a selective and non-ATP-competitive inhibitor of PLK1 with an IC50 of 9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0589