Learn More
TARGETMOL CHEMICALS INC RILAPLADIB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T167515MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rilapladib (SB 659032) is a selective inhibitor of Lipoprotein-Associated Phospholipase A2(Lp-PLA2 IC50 = 230 pM) and an antagonist of the platelet activating factor receptor. purity: 97%
Catalog No. 50-258-1157