Learn More
Cayman Chemical RIlmenIdIn-d4 500ug
SDP
Supplier:  Cayman Chemical 31691500 UG
Encompass_Preferred
An internal standard for the quantification of rilmenidine by GC- or LC-MS
Catalog No. 50-299-2196