Learn More
Medchemexpress LLC Rilmenidine (hemifumarate) | 207572-68-7 | 99.8% | 238.28 g/mol | C10H16N2O · 1/2 C4H4O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100490A10MG
Encompass_Preferred
Rilmenidine hemifumarate is an orally active research compound that selectively agonizes I1 imidazoline receptors and α2-adrenoceptors. It is used in cellular studies to induce autophagy and modulate apoptotic pathways, and is supplied as a high-purity solid for laboratory research.
  • Selective I1 imidazoline receptor and α2-adrenoceptor agonist.
  • Reported to induce autophagy and affect mitochondrial apoptosis pathways in cell models.
  • High-purity solid suitable for research applications.
  • Stable when stored under recommended conditions (solid: -20°C; in solution: -80°C or -20°C as specified).
  • Supplied in small research pack sizes for experimental use.
Catalog No. 50-002-14168