Learn More
Medchemexpress LLC Rilmenidine phosphate | 85409-38-7 | MFCD00875846 | 99.5% | 278.24 g/mol | C10H19N2O5P | 25 MG

Supplier: Medchemexpress LLC HY100490B25MG
Rilmenidine phosphate is a research-grade imidazoline and α2-adrenergic receptor agonist used to investigate antihypertensive mechanisms and receptor pharmacology. Supplied as a high-purity, water-soluble salt, it is intended for in vitro pharmacology, biochemical assays, and receptor-binding studies.
- Selective I1 imidazoline receptor agonist.
- Suitable for in vitro pharmacology and receptor studies.
- High purity (~99.5%).
- Molecular weight 278.24 g/mol.
- Soluble in water (≈83 mg/mL) and DMSO (≈5 mg/mL) with warming or sonication.
- Available in research pack sizes including 25 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.