Learn More
Medchemexpress LLC Riluzole | 1744-22-5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0211R50MG
Encompass_Preferred
Riluzole is an anticonvulsant agent that functions as a use-dependent Na+ channel blocker. It also demonstrates the ability to inhibit GABA uptake, with an IC50 of 43 μM.
  • Acts as an anticonvulsant agent.
  • Functions as a use-dependent Na+ channel blocker.
  • Inhibits GABA uptake.
  • Inhibits peak autaptic IPSCs slightly at 20 μM, while reliably prolonging them.
  • Enhances responses to 2 μM GABA in a strong, concentration-dependent, and readily reversible manner.
  • Exhibits an EC50 of approximately 60 μM for the potentiation of GABA responses.
Catalog No. 50-004-05483