Learn More
Medchemexpress LLC Riluzole hydrochloride | 850608-87-6 | MFCD02262214 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0211A100MG
Encompass_Preferred
Riluzole hydrochloride is an anticonvulsant agent and belongs to the family of use-dependent Na+ channel blocker which can also inhibit GABA uptake with an IC50 of 43 μM.
  • Anticonvulsant agent
  • Use-dependent Na+ channel blocker
  • Inhibits GABA uptake
  • Targets sodium channel
  • IC50 of 43 μM (GABA receptor)
Catalog No. 50-004-05637