Learn More
Medchemexpress LLC Rimantadine hydrochloride | 1501-84-4 | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0338A1G
Encompass_Preferred
Rimantadine hydrochloride (Flumadine hydrochloride) is an orally active inhibitor for M2 protein that blocks hydrogen ion channel activity, preventing virus entry and replication. It exhibits broad-spectrum antiviral activity. It is for research use only and not sold to patients.
  • Inhibits RNA levels of hepatitis A virus (HAV) in Huh7 and IHH cells with IC50 values of 6.7 and 1.6 μg/mL
  • Increases expression of the autophagy marker LC3 and promotes autophagy in HAV-infected Huh7 cells
  • Exhibits anti-infectious activity when combined with Oseltamivir in H3N2-infected mouse models
Catalog No. 50-004-07253