Learn More
Medchemexpress LLC Rimantadine hydrochloride | 1501-84-4 | 98.0% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB0338A10G
Encompass_Preferred
Rimantadine hydrochloride (Flumadine hydrochloride) is an orally active inhibitor for M2 protein. It blocks hydrogen ion channel activity, prevents virus entry and replication, and exhibits broad-spectrum antiviral activity. It is for research use only.
  • Blocks hydrogen ion channel activity.
  • Prevents virus entry and replication.
  • Exhibits broad-spectrum antiviral activity.
  • Inhibits RNA levels of hepatitis A virus (HAV) in Huh7 and IHH cells.
  • Promotes autophagy in HAV-infected Huh7 cells.
  • Shows anti-infectious activity in H3N2-infected mouse models when combined with oseltamivir.
Catalog No. 50-004-06195