Learn More
Medchemexpress LLC Rimantadine (hydrochloride) | 1501-84-4 | 98.0% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB0338A100G
Encompass_Preferred
Rimantadine hydrochloride is an orally active inhibitor for M2 protein, blocking hydrogen ion channel activity, preventing virus entry and replication, and exhibiting broad-spectrum antiviral activity. It is intended for research use only.
  • Inhibits RNA levels of hepatitis A virus (HAV) in Huh7 and IHH cells.
  • Increases LC3 expression and promotes autophagy in HAV-infected Huh7 cells.
  • Exhibits anti-infectious activity in H3N2-infected mouse models, lowering lung virus titer.
Catalog No. 50-004-07134