Learn More
Medchemexpress LLC Rimantadine hydrochloride | 1501-84-4 | 98.0% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB0338A50G
Encompass_Preferred
Rimantadine hydrochloride is an orally active inhibitor for M2 protein. It blocks hydrogen ion channel activity, preventing virus entry and replication, and exhibiting broad-spectrum antiviral activity. It has shown efficacy in inhibiting RNA levels of hepatitis A virus (HAV) in Huh7 and IHH cells and promoting autophagy. In vivo studies also indicate anti-infectious activity in H3N2-infected mouse models.
  • Blocks hydrogen ion channel activity
  • Prevents virus entry and replication
  • Exhibits broad-spectrum antiviral activity
  • Inhibits RNA levels of hepatitis A virus (HAV) in Huh7 and IHH cells
  • Increases expression of autophagy marker LC3
  • Inhibits mTOR phosphorylation
  • Increases lysosome number
  • Promotes autophagy in HAV-infected Huh7 cells
  • Exhibits anti-infectious activity in H3N2-infected mouse models
Catalog No. 50-004-06197