Learn More
Medchemexpress LLC Rimcazole dihydrochloride | 75859-03-9 | 100.0% | 394.38 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10851050MG
Encompass_Preferred
Rimcazole dihydrochloride is a carbazole derivative that functions as a sigma (σ) receptor antagonist. It also exhibits moderate affinity for the dopamine transporter, inhibiting dopamine uptake. This compound has been shown to reduce locomotor activity and sensitization. Rimcazole dihydrochloride has potential applications in cancer research.
  • Functions as a sigma (σ) receptor antagonist.
  • Exhibits moderate affinity for the dopamine transporter.
  • Inhibits dopamine uptake.
  • Reduces locomotor activity and sensitization.
  • Potential applications in cancer research.
Catalog No. 50-003-46991