Learn More
Medchemexpress LLC HY-16046 50mg , Rimiducid CAS:195514-63-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1604650MG
Encompass_Preferred
HY-16046 50mg Rimiducid CAS: 195514-63-7 Rimiducid (AP1903) is a dimerizer agent that acts by cross-linking the FKBP domains. Rimiducid (AP1903) dimerizes the Caspase 9 suicide switch and rapidly induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9763