Learn More
Selleck Chemical LLC Rimiducid S9726-100MG
SDP
Supplier:  Selleck Chemical LLC S9726100MG
Encompass_Preferred
Rimiducid (AP1903) is a chemical protein dimerizer and binds tightly to the target FKBP fusion protein while binding minimally to the abundant natural FKBP Rimiducid elicites potent and dose-dependent apoptosis of cells in culture expressing dimerizer-dependent Fas constructs with an EC50 of 0 1 nM
Catalog No. 50-000-24813