Learn More
Medchemexpress LLC Rimonabant Hydrochloride | 158681-13-1 | 99.3% | C₂₂H₂₂Cl₄N₄O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1413710MM/1ML
Encompass_Preferred
Rimonabant Hydrochloride is a highly potent and selective central cannabinoid receptor (CB1) antagonist with a Ki of 1.8 nM. It also inhibits Mycobacterial membrane protein Large 3 (MMPL3). This product is intended for research use only.
  • Highly potent and selective central cannabinoid receptor (CB1) antagonist
  • Inhibits mycobacterial membrane protein Large 3 (MMPL3)
  • Soluble in DMSO at 33.33 mg/mL (66.63 mM)
  • Store solid at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 1 year or -20°C for 6 months, sealed, away from moisture
  • For research use only
Catalog No. 50-003-53212