Learn More
Selleck Chemical LLC Rimonabant hydrochloride E4858-5MG
SDP
Supplier:  Selleck Chemical LLC E48585MG
Encompass_Preferred
Rimonabant hydrochloride (SR 141716A hydrochloride) is a highly potent and selective antagonist of the central cannabinoid receptor (CB1) with a Ki of 1 8 nM It is an anorectic anti-obesity drug with potential to treat obesity and cardiovascular risk factors
Catalog No. 50-000-42364