Learn More
Medchemexpress LLC Rimonabant (SR141716) | 168273-06-1 | 99.52% | C22H21Cl3N4O | 100 MG

Supplier: Medchemexpress LLC HY141716100MG
Rimonabant is a highly potent, brain-penetrated, and selective central cannabinoid receptor (CB1) antagonist. It also inhibits Mycobacterial membrane protein Large 3 (MMPL3).
- Highly potent, brain-penetrated, and selective central cannabinoid receptor (CB1) antagonist
- Inhibits mycobacterial membrane protein large 3 (MMPL3)
- Ki of 1.8 nM for CB1
- Molecular weight of 463.79
- Appears as a solid
- Color is white to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.