Learn More
ATCC RIN-m5F (ATCC CRL-3660) rat pancreatic beta cell line, frozen vial
Supplier:  ATCC CRL3660
Encompass
RIN-m5F (CRL-3660) is a rat pancreatic beta cell line that produces and secretes insulin and expresses L-dopa decarboxylase. Supplied as frozen cultures, it is used for research into beta cell function and insulin secretion.
  • Derived from rat pancreatic islet beta cells.
  • Produces and secretes insulin and expresses L-dopa decarboxylase.
  • Supplied as frozen vials for recovery and culture.
  • Biosafety level 1; handle using standard laboratory practices.
  • Maintain in RPMI-1640 medium supplemented with 10% fetal bovine serum at 37°C and 5% CO2.
  • Store long term in the vapor phase of liquid nitrogen.
Catalog No. NC3851266