Learn More
Sigma Aldrich Fine Chemicals Biosciences Rink Amide AM resin 100 20
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 8551300025
Encompass
This support comprises the modified Rink amide linker attached to aminomethylpolystyrene and is an ideal tool for the fmoc SPPS of peptide amides. Cleavage from this resin can be effected by a single step treatment with 95% TfA providing peptide amides in high yields and purities.
Catalog No. 50-290-3880